siRNA Details
| Gene Symbol: IGHV5-78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC002792.2 | - | 2 | 506 | 3621362 | |
| X58398.1 | - | 2 | 504 | |||
| X58399.1 | - | 2 | 487 | |||
| Gene Symbol: MIR5195 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_049827.1 | 11 | ||||

IGHV5-78
Genomic Map