siRNA Details
| Gene Symbol: FAM151A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176782.2 | NP_788954.2 | 1 | 162 | ||
| XM_006710599.3 | XP_006710662.1 | 1 | 162 | |||
| GenBank mRNA | AK091901.1 | - | 1 | 191 | ||
| AY358629.1 | - | 1 | 148 | |||
| BC015993.1 | - | 1 | 164 | 4614435 | ||
| BC020874.1 | - | 1 | 160 | 4608844 | ||

FAM151A
Genomic Map