siRNA Details
| Gene Symbol: C2orf48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182626.3 | NP_872432.1 | 3 | 396 | ||
| GenBank mRNA | AK057831.1 | - | 3 | 396 | ||
| BC093999.1 | - | 2 | 138 | 7939844 | ||
| BC094001.1 | - | 2 | 137 | 7939846 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C2orf48| Gene Symbol: C2orf48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182626.3 | NP_872432.1 | 3 | 396 | ||
| GenBank mRNA | AK057831.1 | - | 3 | 396 | ||
| BC093999.1 | - | 2 | 138 | 7939844 | ||
| BC094001.1 | - | 2 | 137 | 7939846 | ||