siRNA Details
| Gene Symbol: WDR18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024100.3 | NP_077005.2 | 1 | 96 | ||
| XM_017027027.1 | XP_016882516.1 | 3 | 622 | |||
| GenBank mRNA | AL560525.3 | - | 1 | 66 | ||
| BC000040.2 | - | 1 | 81 | 3507673 | ||
| BC001648.2 | - | 1 | 76 | 2819452 | ||
| CB991338.1 | - | 123 | ||||

WDR18
Genomic Map