siRNA Details
| Gene Symbol: LCE1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178349.1 | NP_848126.1 | 1 | 941 | ||
| GenBank mRNA | BC031811.1 | - | 1 | 941 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LCE1B| Gene Symbol: LCE1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178349.1 | NP_848126.1 | 1 | 941 | ||
| GenBank mRNA | BC031811.1 | - | 1 | 941 | ||