siRNA Details
| Gene Symbol: PLEKHA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175058.4 | NP_778228.3 | 23 | 4631 | ||
| XM_017017241.1 | XP_016872730.1 | 25 | 4842 | |||
| XM_017017242.1 | XP_016872731.1 | 24 | 4722 | |||
| XM_017017243.1 | XP_016872732.1 | 23 | 4641 | |||
| GenBank mRNA | BC033239.1 | - | 14 | 3511 | 5429651 | |
| BC041452.1 | - | 5 | 1969 | |||
| BC071599.1 | - | 23 | 4627 | 30343461 | ||
| BX648885.1 | - | 2 | 3414 | |||

PLEKHA7
Genomic Map