siRNA Details
| Gene Symbol: SLC51B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178859.3 | NP_849190.2 | 4 | 581 | ||
| XM_005254159.4 | XP_005254216.1 | 4 | 556 | |||
| GenBank mRNA | AK129601.1 | - | 4 | 484 | ||
| AY194242.1 | - | 3 | 290 | |||
| BC103842.1 | - | 3 | 287 | 40002450 | ||
| BC103843.1 | - | 3 | 287 | 40002452 | ||
| BC103844.1 | - | 3 | 287 | 40002453 | ||

SLC51B
Genomic Map