siRNA Details
| Gene Symbol: KRTAP13-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC113536.1 | - | 1 | 64 | 8322588 | |
| BC113538.1 | - | 1 | 64 | 8322590 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRTAP13-1| Gene Symbol: KRTAP13-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC113536.1 | - | 1 | 64 | 8322588 | |
| BC113538.1 | - | 1 | 64 | 8322590 | ||