siRNA Details
Gene Symbol: CCDC136 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201372.1 | NP_001188301.1 | 10 | 1698 | ||
NM_022742.4 | NP_073579.4 | 18 | 4190 | |||
XM_005250534.3 | XP_005250591.1 | 18 | 3867 | |||
XM_005250538.1 | XP_005250595.1 | 16 | 3267 | |||
XM_011516483.1 | XP_011514785.1 | 19 | 3939 | |||
XM_011516484.1 | XP_011514786.1 | 19 | 3758 | |||
XM_011516485.1 | XP_011514787.1 | 19 | 3936 | |||
XM_011516486.1 | XP_011514788.1 | 19 | 3903 | |||
XM_011516489.1 | XP_011514791.1 | 17 | 3339 | |||
XM_017012532.1 | XP_016868021.1 | 18 | 3831 | |||
XM_017012533.1 | XP_016868022.1 | 18 | 4333 | |||
XM_017012534.1 | XP_016868023.1 | 16 | 3231 | |||
XM_017012535.1 | XP_016868024.1 | 13 | 2216 | |||
XM_017012536.1 | XP_016868025.1 | 13 | 2180 | |||
GenBank mRNA | AB058696.1 | - | 18 | 4141 | ||
AK023500.1 | - | 8 | 1840 | |||
AK124447.1 | - | 10 | 1664 | |||
AL133027.1 | - | 14 | 3297 | |||
BC070114.1 | - | 14 | 3820 | |||
BC150331.1 | - | 18 | 4152 | 8860498 | ||
BC152408.1 | - | 18 | 4141 | 8862511 |
