siRNA Details
| Gene Symbol: IFNA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002170.3 | NP_002161.2 | 1 | 346 | ||
| GenBank mRNA | BC104828.1 | - | 1 | 345 | 8143831 | |
| BC104830.1 | - | 1 | 345 | 8143833 | ||
| K01900.1 | - | 1 | 360 | |||
| V00545.1 | - | 1 | 347 | |||
| V00550.1 | - | 1 | 347 | |||

IFNA8
Genomic Map