siRNA Details
| Gene Symbol: KLHL17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198317.2 | NP_938073.1 | 12 | 2399 | ||
| XM_006710600.3 | XP_006710663.1 | 12 | 2952 | |||
| GenBank mRNA | AK127585.1 | - | 11 | 2046 | ||
| AY423763.1 | - | 12 | 2399 | |||
| BC033192.1 | - | 5 | 1088 | |||
| BC105742.1 | - | 9 | 1898 | |||

KLHL17
Genomic Map