siRNA Details
| Gene Symbol: REG1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002909.4 | NP_002900.2 | 6 | 648 | ||
| GenBank mRNA | AK291981.1 | - | 6 | 616 | ||
| BC005350.2 | - | 5 | 592 | 3950349 | ||
| M18963.1 | - | 5 | 600 | |||
| M27190.1 | - | 5 | 627 | |||

REG1A
Genomic Map