siRNA Details
| Gene Symbol: ZNF205 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042428.1 | NP_001035893.1 | 5 | 574 | ||
| NM_001278158.1 | NP_001265087.1 | 5 | 893 | |||
| NM_003456.2 | NP_003447.2 | 5 | 642 | |||
| XM_005255558.2 | XP_005255615.1 | 5 | 609 | |||
| GenBank mRNA | AB672633.1 | - | 5 | 886 | ||
| AF060865.1 | - | 5 | 599 | |||
| AK313009.1 | - | 5 | 631 | |||
| BC002810.2 | - | 5 | 599 | 3635500 | ||

ZNF205
Genomic Map