siRNA Details
| Gene Symbol: FAM151B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_205548.2 | NP_991111.2 | 6 | 1449 | ||
| GenBank mRNA | AY358256.1 | - | 6 | 1449 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM151B| Gene Symbol: FAM151B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_205548.2 | NP_991111.2 | 6 | 1449 | ||
| GenBank mRNA | AY358256.1 | - | 6 | 1449 | ||