siRNA Details
| Gene Symbol: C6orf201 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085401.2 | NP_001078870.1 | 6 | 1482 | ||
| NR_104463.1 | 8 | 2899 | ||||
| NR_104464.1 | 6 | 2245 | ||||
| XM_017010861.1 | XP_016866350.1 | 5 | 1113 | |||
| GenBank mRNA | AF429972.1 | - | 5 | 932 | ||
| BC047663.1 | - | 6 | 1479 | 5266746 | ||
| BC066986.1 | - | 6 | 2234 | 4827732 | ||

C6orf201
Genomic Map