siRNA Details
| Gene Symbol: KRTDAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001244847.1 | NP_001231776.1 | 5 | 403 | ||
| NM_207392.2 | NP_997275.1 | 6 | 445 | |||
| GenBank mRNA | AJ293408.1 | - | 6 | 406 | ||
| AY359007.1 | - | 6 | 368 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRTDAP| Gene Symbol: KRTDAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001244847.1 | NP_001231776.1 | 5 | 403 | ||
| NM_207392.2 | NP_997275.1 | 6 | 445 | |||
| GenBank mRNA | AJ293408.1 | - | 6 | 406 | ||
| AY359007.1 | - | 6 | 368 | |||