siRNA Details
| Gene Symbol: LOC105370980 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011522308.2 | XP_011520610.1 | 1 | 459 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105370980| Gene Symbol: LOC105370980 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011522308.2 | XP_011520610.1 | 1 | 459 | ||