siRNA Details
| Gene Symbol: PTCHD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013732.3 | NP_001013754.3 | 2 | 569 | ||
| NM_207499.2 | NP_997382.2 | 2 | 569 | |||
| XM_011514639.2 | XP_011512941.1 | 2 | 178 | |||
| XM_017010891.1 | XP_016866380.1 | 2 | 709 | |||
| XM_017010892.1 | XP_016866381.1 | 4 | 1753 | |||
| XM_017010893.1 | XP_016866382.1 | 4 | 1579 | |||
| XM_017010894.1 | XP_016866383.1 | 2 | 373 | |||
| XM_017010895.1 | XP_016866384.1 | 2 | 695 | |||
| XM_017010896.1 | XP_016866385.1 | 4 | 1579 | |||
| GenBank mRNA | AK123835.1 | - | 2 | 533 | ||
| AK299301.1 | - | 2 | 569 | |||
| AK300335.1 | - | 2 | 270 | |||
| BC136779.1 | - | 2 | 530 | |||
| DR761295.1 | - | 2 | 243 | |||
| KJ534581.1 | - | 4 | 786 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_028474, BC145294, BC151166.

PTCHD4
Genomic Map