siRNA Details
| Gene Symbol: IHH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002181.3 | NP_002172.2 | 3 | 1153 | ||
| GenBank mRNA | BC034757.1 | - | 3 | 823 | ||
| BC136587.1 | - | 3 | 1153 | 9020577 | ||
| BC136588.1 | - | 3 | 1153 | 9020578 | ||
| L38517.1 | - | 2 | 808 | |||

IHH
Genomic Map