siRNA Details
| Gene Symbol: CFHR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001201550.2 | NP_001188479.1 | 2 | 290 | ||
| NM_001201551.1 | NP_001188480.1 | 2 | 287 | |||
| NM_006684.4 | NP_006675.2 | 2 | 290 | |||
| XM_006711129.3 | XP_006711192.1 | 2 | 229 | |||
| XM_017000110.1 | XP_016855599.1 | 2 | 253 | |||
| XM_017000111.1 | XP_016855600.1 | 2 | 253 | |||
| XM_017000112.1 | XP_016855601.1 | 2 | 253 | |||
| XM_017000113.1 | XP_016855602.1 | 2 | 253 | |||
| GenBank mRNA | AJ640130.2 | - | 2 | 219 | ||
| AK292752.1 | - | 2 | 287 | |||
| X98337.1 | - | 2 | 238 | |||
| Gene Symbol: CFHR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BG618529.1 | - | 2 | 241 | ||

CFHR4
Genomic Map