siRNA Details
| Gene Symbol: PMVK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001323011.1 | NP_001309940.1 | 5 | 564 | ||
| NM_001323012.1 | NP_001309941.1 | 5 | 571 | |||
| NM_006556.3 | NP_006547.1 | 5 | 914 | |||
| GenBank mRNA | AK130234.1 | - | 6 | 802 | ||
| BC006089.1 | - | 5 | 897 | 3604167 | ||
| BC007694.2 | - | 5 | 827 | 3954909 | ||
| L77213.1 | - | 5 | 626 | |||

PMVK
Genomic Map