siRNA Details
| Gene Symbol: Acy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005383.1 | NP_001005383.1 | 8, 9 | 651 | ||
| XM_006243715.2 | XP_006243777.1 | 9, 10 | 953 | |||
| XM_006243716.2 | XP_006243778.1 | 9, 10 | 957 | |||
| GenBank mRNA | AY580164.1 | - | 8, 9 | 651 | ||
| AY580165.1 | - | 8, 9 | 723 | |||
| BC078930.1 | - | 9, 10 | 714 | 7107165 | ||
| FQ232544.1 | - | 9, 10 | 809 | |||

Acy1
Genomic Map