siRNA Details
| Gene Symbol: Rps25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005528.1 | NP_001005528.1 | 3 | 227 | ||
| GenBank mRNA | BC103658.1 | - | 3 | 232 | 7446085 | |
| FQ210084.1 | - | 3 | 245 | |||
| FQ217008.1 | - | 3 | 233 | |||
| FQ217453.1 | - | 3 | 217 | |||
| FQ221194.1 | - | 3 | 248 | |||
| FQ221611.1 | - | 3 | 247 | |||
| FQ221873.1 | - | 3 | 246 | |||
| FQ228420.1 | - | 3 | 248 | |||

Rps25
Genomic Map