siRNA Details
| Gene Symbol: Sorbs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005762.1 | NP_001005762.1 | 17, 18 | 1637 | ||
| XM_006252241.2 | XP_006252303.1 | 18, 19 | 3321 | |||
| XM_006252242.1 | XP_006252304.1 | 17, 18 | 1728 | |||
| XM_006252244.2 | XP_006252306.1 | 16, 17 | 1956 | |||
| XM_006252246.1 | XP_006252308.1 | 17, 18 | 1673 | |||
| XM_006252248.1 | XP_006252310.1 | 5, 6 | 521 | |||
| XM_008770819.1 | XP_008769041.1 | 15, 16 | 3430 | |||
| GenBank mRNA | AF439378.1 | - | 16, 17 | 1453 | ||
| BC083666.1 | - | 17, 18 | 1637 | 7135547 | ||

Sorbs3
Genomic Map