siRNA Details
| Gene Symbol: Atp2b4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005871.1 | NP_001005871.1 | 1 | 180 | ||
| XM_008769446.1 | XP_008767668.1 | 2 | 563 | |||
| XM_008769447.1 | XP_008767669.1 | 2 | 1200 | |||
| XM_008769448.1 | XP_008767670.1 | 2 | 563 | |||
| XM_008769449.1 | XP_008767671.1 | 2 | 564 | |||
| XM_008769450.1 | XP_008767672.1 | 2 | 564 | |||
| GenBank mRNA | U15408.1 | - | 1 | 180 | ||

Atp2b4
Genomic Map