siRNA Details
| Gene Symbol: Clec4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005890.2 | NP_001005890.1 | 3 | 398 | ||
| GenBank mRNA | AY397672.1 | - | 3 | 299 | ||
| AY494063.1 | - | 3 | 299 | |||
| BC098830.1 | - | 3 | 398 | 7378545 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clec4a1| Gene Symbol: Clec4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005890.2 | NP_001005890.1 | 3 | 398 | ||
| GenBank mRNA | AY397672.1 | - | 3 | 299 | ||
| AY494063.1 | - | 3 | 299 | |||
| BC098830.1 | - | 3 | 398 | 7378545 | ||