siRNA Details
| Gene Symbol: Cldnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006955.1 | NP_001006956.1 | 5 | 1568 | ||
| XM_006248204.2 | XP_006248266.1 | 6 | 1761 | |||
| XM_006248205.2 | XP_006248267.1 | 5 | 1710 | |||
| XM_006248206.2 | XP_006248268.1 | 6 | 1612 | |||
| XM_008768633.1 | XP_008766855.1 | 5 | 1552 | |||
| GenBank mRNA | BC083649.1 | - | 5 | 1568 | 7135100 | |

Cldnd1
Genomic Map