siRNA Details
| Gene Symbol: Katnal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006956.1 | NP_001006957.1 | 724 | |||
| XM_006248824.2 | XP_006248886.1 | 1010 | ||||
| GenBank mRNA | BC083673.1 | - | 724 | 7135708 | ||
| Gene Symbol: LOC103690050 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751116.3 | XP_003751164.1 | 6 | 724 | ||
| XM_006248816.2 | XP_006248878.1 | 6 | 1010 | |||
| XM_006248817.2 | XP_006248879.1 | 6 | 743 | |||

Katnal1
Genomic Map