siRNA Details
| Gene Symbol: Kars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006967.1 | NP_001006968.1 | 8 | 1029 | ||
| XM_006255630.1 | XP_006255692.1 | 7 | 901 | |||
| GenBank mRNA | BC083652.1 | - | 8 | 1029 | 7135143 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kars| Gene Symbol: Kars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006967.1 | NP_001006968.1 | 8 | 1029 | ||
| XM_006255630.1 | XP_006255692.1 | 7 | 901 | |||
| GenBank mRNA | BC083652.1 | - | 8 | 1029 | 7135143 | |