siRNA Details
| Gene Symbol: Irf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006969.1 | NP_001006970.1 | 6 | 870 | ||
| XM_006229021.1 | XP_006229083.1 | 6 | 1472 | |||
| XM_006229022.2 | XP_006229084.1 | 6 | 1432 | |||
| XM_006229023.2 | XP_006229085.1 | 6 | 991 | |||
| XM_006229024.2 | XP_006229086.1 | 6 | 859 | |||
| XM_008759337.1 | XP_008757559.1 | 5 | 820 | |||
| GenBank mRNA | BC083776.1 | - | 6 | 870 | 7193483 | |
| FQ218848.1 | - | 6 | 1018 | |||

Irf3
Genomic Map