siRNA Details
| Gene Symbol: Uqcrc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006970.1 | NP_001006971.1 | 14 | 1435 | ||
| GenBank mRNA | BC083610.1 | - | 14 | 1435 | 7133503 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Uqcrc2| Gene Symbol: Uqcrc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006970.1 | NP_001006971.1 | 14 | 1435 | ||
| GenBank mRNA | BC083610.1 | - | 14 | 1435 | 7133503 | |