siRNA Details
| Gene Symbol: Cln3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006971.1 | NP_001006972.1 | 5 | 421 | ||
| XM_006230228.2 | XP_006230290.1 | 6 | 1118 | |||
| XM_008759871.1 | XP_008758093.1 | 6 | 1229 | |||
| XM_008759872.1 | XP_008758094.1 | 7 | 723 | |||
| XM_008759873.1 | XP_008758095.1 | 7 | 798 | |||
| XM_008759874.1 | XP_008758096.1 | 7 | 1035 | |||
| XM_008759875.1 | XP_008758097.1 | 7 | 1175 | |||
| GenBank mRNA | BC083704.1 | - | 5 | 421 | 7190222 | |

Cln3
Genomic Map