siRNA Details
| Gene Symbol: Cln3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006971.1 | NP_001006972.1 | 8 | 662 | ||
| XM_006230228.2 | XP_006230290.1 | 9 | 1359 | |||
| XM_008759871.1 | XP_008758093.1 | 9 | 1470 | |||
| XM_008759872.1 | XP_008758094.1 | 10 | 964 | |||
| XM_008759873.1 | XP_008758095.1 | 10 | 1039 | |||
| XM_008759874.1 | XP_008758096.1 | 10 | 1276 | |||
| XM_008759875.1 | XP_008758097.1 | 10 | 1416 | |||
| GenBank mRNA | BC083704.1 | - | 8 | 662 | 7190222 | |

Cln3
Genomic Map