siRNA Details
| Gene Symbol: Adad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006977.2 | NP_001006978.2 | 3 | 420 | ||
| XM_006232278.2 | XP_006232340.1 | 3 | 604 | |||
| XM_006232279.2 | XP_006232341.1 | 4 | 456 | |||
| GenBank mRNA | BC105906.1 | - | 3 | 486 | 7453283 | |
| CV126304.1 | - | 3 | 451 | |||

Adad1
Genomic Map