siRNA Details
| Gene Symbol: Bcap29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006980.1 | NP_001006981.1 | 7 | 1696 | ||
| XM_006239982.2 | XP_006240044.1 | 8 | 1817 | |||
| GenBank mRNA | BC083576.1 | - | 7 | 1696 | 7113186 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bcap29| Gene Symbol: Bcap29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006980.1 | NP_001006981.1 | 7 | 1696 | ||
| XM_006239982.2 | XP_006240044.1 | 8 | 1817 | |||
| GenBank mRNA | BC083576.1 | - | 7 | 1696 | 7113186 | |