siRNA Details
| Gene Symbol: Dlst | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006981.2 | NP_001006982.2 | 12 | 978 | ||
| XM_006240306.1 | XP_006240368.1 | 10 | 1010 | |||
| XM_008764756.1 | XP_008762978.1 | 11 | 909 | |||
| GenBank mRNA | AB504752.1 | - | 5 | 383 | ||
| BC083858.1 | - | 12 | 964 | 7125734 | ||
| D90401.1 | - | 12 | 929 | |||
| FQ217447.1 | - | 11 | 903 | |||
| FQ217539.1 | - | 11 | 903 | |||

Dlst
Genomic Map