siRNA Details
| Gene Symbol: Vezt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006984.1 | NP_001006985.1 | 12 | 2365 | ||
| XM_006241252.2 | XP_006241314.1 | 12 | 2412 | |||
| GenBank mRNA | BC083840.1 | - | 12 | 2365 | 7114280 | |
| FQ211922.1 | - | 12 | 3285 | |||

Vezt
Genomic Map