siRNA Details
| Gene Symbol: Shisa5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006989.1 | NP_001006990.1 | 4 | 616 | ||
| XM_006243760.1 | XP_006243822.1 | 6 | 948 | |||
| XM_006243761.1 | XP_006243823.1 | 6 | 945 | |||
| XM_006243762.2 | XP_006243824.1 | 4 | 934 | |||
| XM_008766511.1 | XP_008764733.1 | 5 | 646 | |||
| GenBank mRNA | BC083710.1 | - | 4 | 616 | 7190608 | |
| FQ214071.1 | - | 4 | 592 | |||
| FQ231597.1 | - | 6 | 932 | |||

Shisa5
Genomic Map