siRNA Details
| Gene Symbol: CLINT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195555.1 | NP_001182484.1 | 12 | 2342 | ||
| NM_001195556.1 | NP_001182485.1 | 12 | 2319 | |||
| NM_014666.3 | NP_055481.1 | 12 | 2288 | |||
| XM_011534701.2 | XP_011533003.1 | 12 | 2353 | |||
| XM_017010087.1 | XP_016865576.1 | 12 | 2299 | |||
| GenBank mRNA | AK092765.1 | - | 12 | 2289 | ||
| AK128594.1 | - | 2 | 2569 | |||
| AL833548.1 | - | 2 | 1751 | |||
| BC013091.1 | - | 5 | 1153 | |||
| BC030264.1 | - | 4 | 2244 | |||
| D79993.1 | - | 12 | 2184 | |||

CLINT1
Genomic Map