siRNA Details
| Gene Symbol: Igsf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006990.2 | NP_001006991.1 | 4 | 865 | ||
| XM_006248155.2 | XP_006248217.1 | 4 | 730 | |||
| GenBank mRNA | AY325233.1 | - | 3 | 570 | ||
| AY439284.1 | - | 3 | 511 | |||
| BC086342.1 | - | 4 | 571 | |||
| BC097947.1 | - | 4 | 865 | 7373302 | ||

Igsf5
Genomic Map