siRNA Details
| Gene Symbol: Nudt9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006991.1 | NP_001006992.1 | 1, 2 | 205 | ||
| GenBank mRNA | BC083722.1 | - | 1, 2 | 205 | 7190984 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nudt9| Gene Symbol: Nudt9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006991.1 | NP_001006992.1 | 1, 2 | 205 | ||
| GenBank mRNA | BC083722.1 | - | 1, 2 | 205 | 7190984 | |