siRNA Details
| Gene Symbol: Sgcg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006993.1 | NP_001006994.1 | 7 | 738 | ||
| GenBank mRNA | BC083748.1 | - | 7 | 738 | 7192349 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sgcg| Gene Symbol: Sgcg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006993.1 | NP_001006994.1 | 7 | 738 | ||
| GenBank mRNA | BC083748.1 | - | 7 | 738 | 7192349 | |