siRNA Details
| Gene Symbol: Aldh3b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006998.1 | NP_001006999.1 | 10 | 1286 | ||
| XM_006230787.2 | XP_006230849.1 | 11 | 1589 | |||
| XM_008760132.1 | XP_008758354.1 | 10 | 1776 | |||
| XM_008760134.1 | XP_008758356.1 | 10 | 1270 | |||
| GenBank mRNA | BC083850.1 | - | 10 | 1286 | 7122451 | |
| FQ212890.1 | - | 4 | 401 | |||
| Gene Symbol: LOC688778 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760188.1 | XP_008758410.1 | 10 | 1364 | ||

Aldh3b1
Genomic Map