siRNA Details
| Gene Symbol: Pcgf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007000.1 | NP_001007001.1 | 5 | 430 | ||
| GenBank mRNA | AY662670.1 | - | 5 | 430 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcgf1| Gene Symbol: Pcgf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007000.1 | NP_001007001.1 | 5 | 430 | ||
| GenBank mRNA | AY662670.1 | - | 5 | 430 | ||