siRNA Details
Gene Symbol: KIAA0391 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256678.1 | NP_001243607.1 | 2 | 736 | ||
NM_014672.3 | NP_055487.2 | 2 | 736 | |||
XM_005268237.3 | XP_005268294.1 | 2 | 635 | |||
XM_011537409.2 | XP_011535711.1 | 2 | 562 | |||
XM_011537410.2 | XP_011535712.1 | 2 | 620 | |||
XM_017021835.1 | XP_016877324.1 | 2 | 508 | |||
XM_017021836.1 | XP_016877325.1 | 2 | 620 | |||
XM_017021837.1 | XP_016877326.1 | 2 | 619 | |||
GenBank mRNA | AB002389.1 | - | 2 | 486 | ||
AK304066.1 | - | 2 | 493 | |||
BC032221.1 | - | 2 | 487 | 5206545 | ||
BC044580.1 | - | 2 | 505 | 4822711 | ||
BX161394.1 | - | 2 | 468 |
