siRNA Details
| Gene Symbol: Mxra8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007002.1 | NP_001007003.1 | 8 | 2064 | ||
| XM_006239574.1 | XP_006239636.1 | 10 | 2289 | |||
| XM_006239575.1 | XP_006239637.1 | 10 | 2280 | |||
| GenBank mRNA | BC083849.1 | - | 8 | 2064 | 7122386 | |

Mxra8
Genomic Map