siRNA Details
| Gene Symbol: Arhgdia | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007005.1 | NP_001007006.1 | 6 | 780 | ||
| XM_006247897.2 | XP_006247959.1 | 6 | 914 | |||
| XM_006247898.2 | XP_006247960.1 | 6 | 863 | |||
| GenBank mRNA | BC083817.1 | - | 6 | 780 | 7110945 | |

Arhgdia
Genomic Map