siRNA Details
| Gene Symbol: Dusp13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007006.1 | NP_001007007.1 | 556 | |||
| NM_001162408.1 | NP_001155880.1 | 1027 | ||||
| XM_008770542.1 | XP_008768764.1 | 4 | 1188 | |||
| XM_008770543.1 | XP_008768765.1 | 1386 | ||||
| XM_008770544.1 | XP_008768766.1 | 3 | 984 | |||
| GenBank mRNA | BC083646.1 | - | 556 | 7135038 | ||
| BC166532.1 | - | 1027 | 7190084 | |||
| Gene Symbol: LOC100909469 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751425.3 | XP_003751473.2 | 4 | 594 | ||
| XM_003751426.3 | XP_003751474.1 | 2 | 1386 | |||
| XM_008770474.1 | XP_008768696.1 | 4 | 1188 | |||
| XM_008770475.1 | XP_008768697.1 | 3 | 984 | |||

Dusp13
Genomic Map