siRNA Details
| Gene Symbol: EMC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK222615.1 | - | 4 | 312 | ||
| AK290974.1 | - | 4 | 298 | |||
| BC020753.1 | - | 4 | 310 | 4809693 | ||
| BC021667.1 | - | 4 | 327 | 3682913 | ||
| CR457402.1 | - | 4 | 288 | |||
| D14659.1 | - | 4 | 294 | |||

EMC2
Genomic Map