siRNA Details
| Gene Symbol: Zpbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007011.1 | NP_001007012.1 | 8 | 1108 | ||
| XM_006247384.2 | XP_006247446.1 | 8 | 2238 | |||
| XM_006247385.2 | XP_006247447.1 | 8 | 2165 | |||
| XM_006247387.2 | XP_006247449.1 | 7 | 2112 | |||
| GenBank mRNA | AY251605.1 | - | 8 | 1100 | ||
| AY251606.1 | - | 7 | 1034 | |||
| BC082007.1 | - | 8 | 1108 | 7131668 | ||

Zpbp2
Genomic Map